H-ORN(Z)-OH 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H62435.03 H62435.06 H62435.14
H62435.03EA 31.1 EUR
H62435.03 H62435.06 H62435.14
H-ORN(Z)-OH 98%
H-ORN(Z)-OH

Purity: 98% CAS No.: 3304-51-6 Molecular Formula: C13H18N2O4 Formula Weight: 266.29 Physical Form: Solid MDL No.: MFCD00038200

Formule: C₁₃H₁₈N₂O₄
Smeltpunt: 248…252 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00037220
CAS nummer: 3304-51-6
EINECS: 221-980-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR