L-Alanine-4-nitroanilide hydrochloride 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H63393.MD H63393.03 H63393.06
H63393.MDEA 47.9 EUR
H63393.MD H63393.03 H63393.06
L-Alanine-4-nitroanilide hydrochloride 98%
L-Alanine-4-nitroanilide hydrochloride

Purity: 98% CAS No.: 31796-55-1 Molecular Formula: C9H11N3O3.HCl Formula Weight: 245.66 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: Pale yellow Beilstein Reference: 6303731 MDL No.: MFCD00039088

Formule: C₉H₁₂ClN₃O₃
MDL Nummer: MFCD00039088
CAS nummer: 31796-55-1
EINECS: 250-811-0

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR