3-Nitro-D-phenylalanine 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H63483.MD H63483.03
H63483.MDEA 70.7 EUR
H63483.MD H63483.03
3-Nitro-D-phenylalanine 95%
3-Nitro-D-phenylalanine

Purity: 95% CAS No.: 169530-97-6 Molecular Formula: C9H10N2O4 Formula Weight: 210.19 Storage Temperature: -30°C to -10°C Physical Form: Solid Appearance: Off-white to pale yellow MDL No.: MFCD04117829

Formule: C₉H₁₀N₂O₄
MDL Nummer: MFCD04117829
CAS nummer: 169530-97-6

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR