Fmoc-Asp(OAll)-OH 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H63585.06 H63585.14
H63585.06EA 100 EUR
H63585.06 H63585.14
Fmoc-Asp(OAll)-OH 98%
Fmoc-Asp(OAll)-OH

Purity: 98% CAS No.: 146982-24-3 Molecular Formula: C22H21NO6 Formula Weight: 395.41 Storage Temperature: 2°C to 8°C Physical Form: Solid Melting Point: 111-115° Beilstein Reference: 6670219 Optical Rotation: -27°(c=1 in DMF) MDL No.: MFCD00190874

Formule: C₂₂H₂₁NO₆
Smeltpunt: 111…115 °C
MDL Nummer: MFCD00190874
CAS nummer: 146982-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR