Nα-(tert-Butoxycarbonyl)-L-arginine hydrochloride 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H63611.06 H63611.14
H63611.06EA 35.8 EUR
H63611.06 H63611.14
Nα-(tert-Butoxycarbonyl)-L-arginine hydrochloride 98%
Nalpha-Boc-L-arginine hydrochloride

Purity: 98% CAS No.: 35897-34-8 Molecular Formula: C11H22N4O4.HCl Formula Weight: 310.78 Physical Form: Powder Appearance: White Beilstein Reference: 3658939 MDL No.: MFCD00063594

Formule: C₁₁H₂₂N₄O₄·HCl
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00063594
CAS nummer: 35897-34-8

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR