L-Proline-tert-butyl ester 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

H63657.03 H63657.06
H63657.03EA 54.8 EUR
H63657.03 H63657.06
L-Proline-tert-butyl ester 98%
L-Proline-tert-butyl ester

Purity: 98% CAS No.: 2812-46-6 Molecular Formula: C9H17NO2 Formula Weight: 171.24 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Colourless MDL No.: MFCD00037879

Formule: C₉H₁₇NO₂
MDL Nummer: MFCD00037879
CAS nummer: 2812-46-6
EINECS: 220-558-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR