5-Bromo-2-chloro-3-methylpyridine 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H64109.06 H64109.14
H64109.06EA 43.8 EUR
H64109.06 H64109.14
5-Bromo-2-chloro-3-methylpyridine 98%
5-Bromo-2-chloro-3-methylpyridine

Purity: 98% CAS No.: 29241-60-9 Molecular Formula: C6H5BrClN Formula Weight: 206.47 Physical Form: Solid Melting Point: 40-44° Beilstein Reference: 1562796 MDL No.: MFCD03095093

Formule: C₆H₅BrClN
Smeltpunt: 40…44 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03095093
CAS nummer: 29241-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR