3-Carboxyphenylboronic acid pinacol ester 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H64195.06 H64195.14
H64195.06EA 63.4 EUR
H64195.06 H64195.14
3-Carboxyphenylboronic acid pinacol ester 97%
3-Carboxyphenylboronic acid pinacol ester

Purity: 97% CAS No.: 269409-73-6 Molecular Formula: C13H17BO4 Formula Weight: 248.09 Physical Form: Powder Appearance: White Melting Point: 206-210° dec. MDL No.: MFCD03411930

Formule: C₁₃H₁₇BO₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03411930
CAS nummer: 269409-73-6

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR