2-Bromo-5-hydroxybenzaldehyde 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

H64884.06 H64884.14 H64884.22
H64884.06EA 49.7 EUR
H64884.06 H64884.14 H64884.22
2-Bromo-5-hydroxybenzaldehyde 95%
2-Bromo-5-hydroxybenzaldehyde

Purity: 95% CAS No.: 2973-80-0 Molecular Formula: C7H5BrO2 Formula Weight: 201.02 Physical Form: Solid Melting Point: 130-135° Beilstein Reference: 2079921 UN No.: UN3077 MDL No.: MFCD03033065

Formule: C₇H₅BrO₂
MW: 201.02 g/mol
Smeltpunt: 130…135 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD03033065
CAS nummer: 2973-80-0
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR