1-tert-Butylpiperazine 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

H66497.03 H66497.06
H66497.03EA 55.4 EUR
H66497.03 H66497.06
1-tert-Butylpiperazine 97%
1-tert-Butylpiperazine

Purity: 97% CAS No.: 38216-72-7 Molecular Formula: C8H18N2 Formula Weight: 142.24 Physical Form: Low melting solid Sensitivity: Light, Air and Moisture Sensitive Appearance: Colourless to yellow Melting Point: 32-35° Boiling Point: 85-87°/22mm UN No.: UN3259 MDL No.: MFCD00049359

Formule: C₈H₁₈N₂
Kookpunt: 85…87 °C (22 mmHg)
Smeltpunt: 32…35 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00049359
CAS nummer: 38216-72-7
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR