Potassium iodide

Leverancier: Hach
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Dipotassium diiodide, Kali iodide

16726H
HACH16726HEA 94.8 EUR
HACH16726H
Potassium iodide
Potassium iodide
Formule: Kl
Kookpunt: 1330 °C (1013 hPa)
Smeltpunt: 686 °C
Dichtheid: 3,13 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011405
CAS nummer: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR