Multiparameter test kits, model HA-62
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
183700 183701 183702
HACH183700EA 379 EUR
HACH183700 HACH183701 HACH183702
Multiparameter test kits, model HA-62
Milieutestkits
A multi-parameter test kit for hardness, iron and pH, which provides an easy-to-use, cost effective way to determine the quality of water.
  • Popular tests in one rugged kit
  • Uses color disc and drop count titration methods
Order Now
Specificaties
Environmentally Preferable