Lauryl tryptose broth tubes
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2162315
HACH2162315EA 61.2 EUR
HACH2162315
Lauryl tryptose broth tubes
Media voor microbiologie Geprepareerde media voor microbiologie
Lauryl tryptose broth tubes, single strength, selectivity: total coliform bacteria (presumptive).
  • Save time and labour costs with ready to use MPN tubes, with minimal training requirements