Test kits, general boiler water, drop count titration AL-94
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
222403
HACH222403EA 531 EUR
HACH222403
Test kits, general boiler water, drop count titration AL-94
Milieutestkits
This kit is used for controlling boiler and cooling water treatment and checking corrosion using the drop count test which is a simple volumetric/ titration method with colour indication. It is an economical solution for the small boiler operator who needs to monitor alkalinity (5 to 400 mg/L), chloride (5 to 400 mg/L), hardness (0 to 2500 mg/L), iron (0 to 4 mg/L), and pH (4 to 10).
  • Simple, accurate methods
  • All-inclusive, the reagents are supplied with the kit
  • Application specific
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Leveringsinformatie: Supplied with ten reagents, one measuring tube, two stoppers, one colour comparator box, mixing bottle, three colour discs, instructions, and carrying case.