Saltwater aquaculture test kit, FF-3
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
243003
HACH243003EA 1604 EUR
HACH243003
Saltwater aquaculture test kit, FF-3
Milieutestkits
Test kit used to measures 10 critical parameters with digital titrator, drop count, and colour discs. It is suitable for use in saltwater aquaculture applications.
Leveringsinformatie: Includes twenty four reagents, one flask, clippers, one graduated cylinder, one colour comparator box, three colour discs, one vial, one delivery tube, one syringe, two plastic tubes, one BOD bottle, one thermometer, one digital titrator, a manual, and a carrying case.