Conductivity and TDS standards, natural water
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2974826 2974849 2974549 2974649
HACH2974826EA 15.2 EUR
HACH2974826 HACH2974849 HACH2974549 HACH2974649
Conductivity and TDS standards, natural water
Standaarden Geleidbaarheidsstandaarden
Natural Water Standard Solution:40% sodium sulphate/40% sodium bicarbonate/20% sodium chloride.