Conductivity cells, Radiometer Analytical
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
B60E100 B15B001 E61M013 E61M008 E61M009 E61M015 E61M010 E61M014 E61M022 E61M011 E61M012 E61M016
HACHB60E100EA 454 EUR
HACHB60E100 HACHB15B001 HACHE61M013 HACHE61M008 HACHE61M009 HACHE61M015 HACHE61M010 HACHE61M014 HACHE61M022 HACHE61M011 HACHE61M012 HACHE61M016
Conductivity cells, Radiometer Analytical
Elektroden Geleidbaarheidselektroden
Range of conductivity cells for a wide variety of applications.
  • Two-pole cells: traditional design based on two plates of platinum; ideal for routine measurement of conductivity and for use with a sample changer due to the easy rinsing
  • three-pole cells: three platinum rings which facilitate optimal shielding during measurement
  • four-pole cells: four platinum rings ensure accurate results over several decades of conductivity with a single cell using just one calibration
  • four-pole cells are particularly recommended when performing high conductivity measurements