Test kits, photometric, nickel acid baths, LCK cuvette tests
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
LCK237
HACHLCK237EA 155 EUR
HACHLCK237
Test kits, photometric, nickel acid baths, LCK cuvette tests
Milieutestkits
Cuvette tests for acidic nickel plating baths, nickel sulphate and nickel sulphamate baths applications.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
Shelf life: 12 months from production date
Storage conditions: 15 to 25 °C