Multi-parameter electrode, sensION+ 5059
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
LZW5059970002
HACHLZW5059970002EA 534 EUR
HACHLZW5059970002
Multi-parameter electrode, sensION+ 5059
Elektroden Multiparameter elektroden
sensION+ 5059 is a combination multi-parameter electrode for determination of pH, conductivity and temperature. It has a plastic body, non-refillable gel reference electrolyte and built-in temperature sensor.
  • Determine multiple parameters in the field with one standard sized electrode
  • Low maintenance design for a variety of applications
  • Ultra portable but protected against harsh field conditions
  • Heavy-duty electrode handle design optimised for field calibration and storage
  • Glass (pH); platinum (conductivity); and Pt1000 (temperature) sensors