pH buffers, DuraCal
Leverancier: HAMILTON BONADUZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
238917 238918 238919 238279 238277 238319 238275 238273 238317 238318 238321 238194 238187 238188 238278 238332 238333
HAMI238917EA 80 EUR
HAMI238917 HAMI238918 HAMI238919 HAMI238279 HAMI238277 HAMI238319 HAMI238275 HAMI238273 HAMI238317 HAMI238318 HAMI238321 HAMI238194 HAMI238187 HAMI238188 HAMI238278 HAMI238332 HAMI238333
pH buffers, DuraCal
Buffers pH-buffers
For full information concerning health and safety data please contact Avantor or Hamilton directly.
  • Label includes expiration date, batch number and actual value
  • Traceable to NIST and PTB, certificates available to download
  • Stable solutions
  • Certified pH value from a DAkkS laboratory accredited for pH measurement