Metal hub needles
Leverancier: HAMILTON BONADUZ
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
90018 90019 90020 90021 90022 90023 90024 90025 90026 90027 90031 90032 90033 90038 90039 91038 91039 91022 7751-12 7748-09 91033 7748-03 7748-08 91023 7729-07 7747-01 7748-02 7748-12 7748-14 7751-07 7751-17 91025 7748-17 7748-18 7748-06 7729-10 91024 91014 7729-03 7748-07 91026 91017 91018 91020 91021 7729-02 7729-04 7729-05 7729-06 7729-08 7729-09 7729-01 7748-16 7748-04 7748-13 7749-05 7748-11 7748-19 7751-14 7749-02 7751-08 7751-09 7751-11 7751-13 7751-15 7751-16 7751-18 7751-19 7748-10 7749-01 7749-03 7749-04 7748-15 7748-05 7751-01 7747-02 7751-02 7751-03 7751-04 7751-05 7751-06 7751-10 7751-20 7751-21 7748-01 7748-20 91019 7730-02 91032 91015
HAMI90018EA 61.7 EUR
HAMI90018 HAMI90019 HAMI90020 HAMI90021 613-0099 HAMI90023 HAMI90024 HAMI90025 HAMI90026 HAMI90027 HAMI90031 HAMI90032 HAMI90033 549-1241 549-1242 HAMI91038 HAMI91039 HAMI91022 HAMI7751-12 HAMI7748-09 HAMI91033 HAMI7748-03 HAMI7748-08 HAMI91023 HAMI7729-07 HAMI7747-01 HAMI7748-02 HAMI7748-12 HAMI7748-14 HAMI7751-07 HAMI7751-17 HAMI91025 HAMI7748-17 HAMI7748-18 HAMI7748-06 HAMI7729-10 HAMI91024 HAMI91014 HAMI7729-03 HAMI7748-07 HAMI91026 HAMI91017 HAMI91018 HAMI91020 HAMI91021 HAMI7729-02 HAMI7729-04 HAMI7729-05 HAMI7729-06 HAMI7729-08 HAMI7729-09 HAMI7729-01 HAMI7748-16 HAMI7748-04 HAMI7748-13 HAMI7749-05 HAMI7748-11 HAMI7748-19 HAMI7751-14 HAMI7749-02 HAMI7751-08 HAMI7751-09 HAMI7751-11 HAMI7751-13 HAMI7751-15 HAMI7751-16 HAMI7751-18 HAMI7751-19 HAMI7748-10 HAMI7749-01 HAMI7749-03 HAMI7749-04 HAMI7748-15 HAMI7748-05 HAMI7751-01 HAMI7747-02 HAMI7751-02 HAMI7751-03 HAMI7751-04 HAMI7751-05 HAMI7751-06 HAMI7751-10 HAMI7751-20 HAMI7751-21 HAMI7748-01 HAMI7748-20 HAMI91019 HAMI7730-02 HAMI91032 HAMI91015
Metal hub needles
Injectienaalden
Metal hub (nickel plated brass) needles can be used with TLL syringes and LT or TLL connectors. The needles are 304 stainless steel. Custom needle lengths and point styles are available.