Universal tongs, type Hünebeck, stainless steel
Leverancier: HAMMACHER KARL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HWN313-19 HWN309-25
HAMMHWN313-19EA 107 EUR
HAMMHWN313-19 HAMMHWN309-25
Universal tongs, type Hünebeck, stainless steel
Tangen
Universal holding tongs made from corrosion resistant WIRONIT steel (18/12 stainless steel with 18% chrome and 10 to 12% nickel).
Order Now
Specificaties
Environmentally Preferable