Kroonstopwatch, ABS
Leverancier: HANHART
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
111.0117-00
HANH111.0117-00EA 281 EUR
HANH111.0117-00
Kroonstopwatch, ABS
Timers Analoge stopwatches
Analogue stopwatch with functional ABS plastic case and pendant ring.
  • Very lightweight and easy to handle
  • Pin lever, 1 jewel
  • 1/5 seconds, 30 minutes