pH Buffer solutions in bottles

Leverancier: Hanna
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

HI7004L HI7007L
HANNHI7004LEA 25.6 EUR
HANNHI7004L HANNHI7007L
pH Buffer solutions in bottles
Buffers pH-buffers

Plastic bottle with a tamper-proof seal.


  • Color coded label


Traceable to NIST standard reference materials.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR