Peha-taft® Latex Powderfree Gloves
Leverancier: HARTMANN PAUL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
942595 9425894 9425904 9425914 9425924 9425934 9425944 9425954 9425964
HART942595EA 98.9 EUR
HART942595 HART9425894 HART9425904 HART9425914 HART9425924 HART9425934 HART9425944 HART9425954 HART9425964
Peha-taft® Latex Powderfree Gloves
Handschoenen
Powder-free disposable surgical gloves made of thin and particularly soft latex, particularly easy to put on thanks to the special polymer inner coating, with a special micro-roughened surface for optimal grip, very good tactile sensitivity, highly elastic and tear-resistant, fatigue-free fit thanks to the fully anatomical fit.
  • Sophisticated
  • Refined and smooth without being slippery
  • Sterile
  • sealed in pairs with ozone protection
Peha-taft® latex is a medical product in accordance with EN 455 and personal protective equipment in accordance with EN 374,More Product Information