Equilibrium DIALYZER™ plate for Single Use 96-Well Equilibrium DIALYZER™
Leverancier: HARVARD APPARATUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
74-2331
HARV74-2331EA 1690 EUR
HARV74-2331
Equilibrium DIALYZER™ plate for Single Use 96-Well Equilibrium DIALYZER™
Microplaten HTS-microplaten
The Single Use 96-Well Equilibrium DIALYZER™ is designed for high throughput protein interaction studies.
  • Each well in this system has a separate membrane, eliminating possibility of cross-contamination
  • Excellent sample recovery
  • Wells are sealed with 8-cap strips (not included)