Coated wire racks
Leverancier: HEATHROW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
120089 120759 120760 120761 120763 120764 120765 120766 120767 120768 120769 120770 120771 120772
HEAT120089EA 56.7 EUR
HEAT120089 HEAT120759 HEAT120760 HEAT120761 HEAT120763 HEAT120764 HEAT120765 HEAT120766 HEAT120767 HEAT120768 HEAT120769 HEAT120770 HEAT120771 HEAT120772
Coated wire racks
Rekken Tube Racks
These HDPE coated wire racks are chemically resistant and suitable for all brands and types of conical, round or flat-bottom, or plastic tubes and bottles.
  • Racks can be used between temperatures of −40 and 121°C
  • Square wells and three deck levels help keep tubes secure
  • Autoclavable
  • Slotted bottom support
These are durable for transport or sample handling.
Order Now
Specificaties
Environmentally Preferable