Mini coolers, True North®
Leverancier: HEATHROW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HS120072 HS120073 HS120074
HEATHS120072EA 599 EUR
HEATHS120072 HEATHS120073 HEATHS120074
Mini coolers, True North®
Dozen Koelboxen en koeltassen
Protect temperature-sensitive samples at the bench or in the freezer.
  • Polycarbonate construction with rubber non-slip feet
  • Gridded and keyed one-position lids for easy sample identification
Ideal for keeping PCR® reagents, enzymes, and other biochemical reagents cool while working at the bench.
Filled with insulating gel, the mini coolers can also be used for controlled sample freezing and for longer temperature retention samples.
Coolers HEATHS120072 and HEATHS120075 are filled with a non-toxic self-insulating gel.
Bestelinformatie: VersionsMore Product Information