Multi-function stopwatch
Leverancier: HEATHROW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HS24645
HEATHS24645EA 65.5 EUR
HEATHS24645
Multi-function stopwatch
Timers Digitale stopwatches
Timing capacity: 60 minutes in ¹/₁₀₀ second increments.
  • Lightweight, dependable, accurate and easy to use
  • Large ¹/₄" display
  • Functions: split, hourly chime, calendar, alarm
Leveringsinformatie: Supplied with neck cord and one LR44 (1,5 V) battery.