Racks for upright freezers
Leverancier: HEATHROW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
HS28622
HEATHS28622EA 390 EUR
HEATHS28622
Racks for upright freezers
Rekken Rekken voor cryogene dozen
Stainless steel.
For upright freezers and used for 15 and 50 ml tube box storage. Corrosion-resistant construction with (2×3) rack spaces.