Electronic cell counters

Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

40345015 40345010
HECH40345015EA 891 EUR
HECH40345015 233-5394
Electronic cell counters
Celtellers Automatische celtellers
The electronic memory devices specify blood counting.

  • Power supply operation
  • Rechargeable battery operation
  • Software (download) with computer analysis
  • Big TFT color display
  • Storage of last counting on device

The microscope image can be observed permanently, the numbers can be typed in easily.

Colour: Blue

Leveringsinformatie: Every device comes with an instruction manual.
Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR