Graduated pipettes, conformity certified, partial delivery
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
40545730 40545736
HECH40545730EA 107 EUR
HECH40545730 HECH40545736
Graduated pipettes, conformity certified, partial delivery
Pipetten Gegradueerde pipetten
Glass.
  • Enzyme test pipettes with long, tapered, sturdy tip
  • Precision type, for partial delivery (not calibrated to tip), calibrated to deliver (Ex),
  • Zero at top, marked main points
  • Tolerance indicated on each pipette, amber graduations