Centrifuge tubes, conical, without rim
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
40939001 40939002 40939302
HECH40939001EA 360 EUR
HECH40939001 HECH40939002 HECH40939302
Centrifuge tubes, conical, without rim
Buisjes Centrifugebuisjes
AR®-Glass (soda glass) or DURAN®, borosilicate glass.
  • Ffused white graduation
  • Good chemical resistance
Conical bottom, rimless.