Mounting medium for histology/microscopy, Histokitt, Assistent®

Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

41025010
HECH41025010EA 62.5 EUR
HECH41025010
Mounting medium for histology/microscopy, Histokitt, Assistent®
Kleurstoffen Microscopy and staining reagents

Mounting medium for histology/microscopy.


  • Light-resistant
  • Neutral
  • Colourless
  • Does not affect colour of the specimen
  • Soluble e.g. In benzene, chloroform, dioxane, xylene
  • In UN certified tinplate bottle


Hardening time 20 minutes, cold-resistant down to -17 °C (+1,4 °F).

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR