Analogue stopwatch
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
41978010
HECH41978010EA 297 EUR
HECH41978010
Analogue stopwatch
Timers Analoge stopwatches
Solid metal stopwatch with 7 jewels.
  • Graduation 0 to 60 in 1/5 seconds and 1/100 minutes
  • Timing capacity: Up to 30 minutes
  • Impact, dust and water resistant