Measuring cylinders, tall form, class B, soda glass
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
42200030 42200048 42200060 42200036 42200042 42200024 42200066 42200018 42200006 42200012
612-9299EA 8.2 EUR
612-9299 612-9449 HECH42200060 HECH42200036 HECH42200042 HECH42200024 HECH42200066 HECH42200018 HECH42200006 HECH42200012
Measuring cylinders, tall form, class B, soda glass
Cilinders Maatcilinders
Soda glass, tall form, class B.
  • Calibrated to contain (marked 'In')
  • Round base, spout
  • Fused amber short line graduation
Certificaten: DIN EN ISO 4788