Pycnometer
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
42571024 42570008 42570016 42570024 42570032 42570040 42571008 42571308
HECH42571024EA 61.1 EUR
HECH42571024 HECH42570008 HECH42570016 HECH42570024 HECH42570032 HECH42570040 HECH42571008 HECH42571308
Pycnometer
Pyknometers
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
  • With ground-in stopper
  • Good chemical resistance
  • Good temperature resistance
Certificaten: DIN ISO 3507
Order Now
Specificaties
Environmentally Preferable