Pycnometer with thermometer, adjusted
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
42572324 42572332 42572340 42572016 42572024 42572032 42572040
HECH42572324EA 124 EUR
HECH42572324 HECH42572332 HECH42572340 HECH42572016 HECH42572024 HECH42572032 HECH42572040
Pycnometer with thermometer, adjusted
Pyknometers
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
  • With ground-in capillary stopper
  • Very good chemical resistance
  • Very good temperature resistance
It includes pycnometer bottle and ground-in thermometer joint size 10/19 with blue filling.
Certificaten: DIN ISO 3507