Weighing bottles, glass
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
44250002 44250004 44250006 44250001 44250003 44250005 44251010 44251011 44251012 44251013 44251014 44251016
611-0098EA 19.9 EUR
611-0098 611-0099 611-0100 HECH44250001 HECH44250003 HECH44250005 HECH44251010 HECH44251011 HECH44251012 HECH44251013 HECH44251014 HECH44251016
Weighing bottles, glass
Flessen Weegflessen
Made of borosilicate glass.
  • Excellent chemical resistance
  • Excellent temperature resistance
With interchangeable lid and frosted label.