Imhoff Cone without Stopcock
Leverancier: GLASWARENFABRIK KARL HECHT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
44275010
HECH44275010EA 164 EUR
HECH44275010
Imhoff Cone without Stopcock
Trechters Sedimentatiekegels
Made from borosilicate 3.3 glass.
  • Very good chemical resistance
  • Excellent thermal resistance
  • With frosted label and scale
  • Fused white graduation
The sedimentation cone with graduation according to Imhoff is used for the quantification of sediments from a suspension.
Certificaten: Produced according to DIN 12672.