Magnetic Stirrers, Hei-PLATE Mix 'n' Heat Ultimate
Leverancier: Heidolph Instruments GmbH & Co.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
506-41100-00 506-41151-00 506-42100-00 506-41158-00 506-41150-00
HEID506-41100-00EA 1730 EUR
HEID506-41100-00 HEID506-41151-00 HEID506-42100-00 HEID506-41158-00 HEID506-41150-00
Magnetic Stirrers, Hei-PLATE Mix 'n' Heat Ultimate
Roerders Magnetische roerders
Hei-PLATE Mix 'n' Heat Ultimate is equipped with numerous interfaces for automated laboratory processes. The RS232 and USB B ports allow a control via our integrated Hei-PROCESS solution.
The USB A interface at the front allows data logging via a USB stick. For more comfort and safety, you can connect the magnetic stirrer with the electric Heidolph lab jack via the USB C interface. There are three heating modes for temperature control: FAST for a quick heating, as well as PRECISE and PRECISE+ for the exact control of your target temperature without overshoot - this is how sensitiveMore Product Information