Rotary Evaporators, Hei-VAP Expert
Leverancier: Heidolph Instruments GmbH & Co.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
573-01100-00 573-01300-00 573-01305-00 573-01500-00 573-01600-00 573-01110-00 573-01310-00 573-01315-00 573-01510-00 573-01610-00 574-01100-00 574-01300-00 574-01305-00 574-01500-00 574-01600-00 574-01110-00 574-01310-00 574-01315-00 574-01510-00 574-01610-00
HEID573-01100-00EA 4740 EUR
HEID573-01100-00 HEID573-01300-00 HEID573-01305-00 HEID573-01500-00 HEID573-01600-00 HEID573-01110-00 HEID573-01310-00 HEID573-01315-00 HEID573-01510-00 HEID573-01610-00 HEID574-01100-00 HEID574-01300-00 HEID574-01305-00 HEID574-01500-00 HEID574-01600-00 HEID574-01110-00 HEID574-01310-00 HEID574-01315-00 HEID574-01510-00 HEID574-01610-00
Rotary Evaporators, Hei-VAP Expert
Verdampers Rotatieverdampers
Precise setting of rotation speed and heating bath temperature on the digital touch display or for quick access during an active process directly via the knobs on the operating panel.
  • Central operating panel with 5" touch display
  • Clear display of all values (Basic and Advanced modes)
  • With timer, limitation of max. temperature, multilingual menu and text display for error messages
  • LED ring lights and pictogram to display the activity even from a distance
  • USB and Micro SD interfaces
  • Optional display of vapour temperature
  • Overtemperature protection: Cut-off
More Product Information