Hei-VAP Ultimate Control Rotary Evaporators
Leverancier: Heidolph Instruments GmbH & Co.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
575-02100-00 575-02300-00 575-02305-00 575-02500-00 575-02600-00 575-02110-00 575-02310-00 575-02315-00 575-02510-00 575-02610-00 576-02100-00 576-02300-00 576-02305-00 576-02500-00 576-02600-00 576-02110-00 576-02310-00 576-02315-00 576-02510-00 576-02610-00 576-26305-00 576-27305-00
HEID575-02100-00EA 6040 EUR
HEID575-02100-00 HEID575-02300-00 HEID575-02305-00 HEID575-02500-00 HEID575-02600-00 HEID575-02110-00 HEID575-02310-00 HEID575-02315-00 HEID575-02510-00 HEID575-02610-00 HEID576-02100-00 HEID576-02300-00 HEID576-02305-00 HEID576-02500-00 HEID576-02600-00 HEID576-02110-00 HEID576-02310-00 HEID576-02315-00 HEID576-02510-00 HEID576-02610-00 HEID576-26305-00 HEID576-27305-00
Hei-VAP Ultimate Control Rotary Evaporators
Verdampers Rotatieverdampers
Adds numerous smart features to the Hei-VAP Ultimate, as well as an overview of all parameters and the current process.
  • 7" touch display for controlling the entire system including the vacuum and the cooling temperature
  • Motor lift model with transparent plastic coated G1 diagonal glassware
  • For precise monitoring, the pictogram on the display also symbolizes active functions
  • Separate knobs for quick access: the right one for direct vacuum control, the left one for the rotation
  • Setting of values either via the touch display or the separate quick access knobs
More Product Information