Rekken voor vrieskisten
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
398187 398185 398186 398184
HERA398187EA 132 EUR
HERA398187 HERA398185 HERA398186 HERA398184
Rekken voor vrieskisten
Rekken Rekken voor cryogene dozen
Use Thermo Scientific™ Chest freezer racks with TDE Series ultra-low temperature chest freezers as well as legacy Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-low temperature chest freezers.