Rekken voor diepvrieskasten met ultralage temperatuur van Revco
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
398331 398324 398332 398330 398329 398328 398327 398325 398326
HERA398331EA 222 EUR
HERA398331 HERA398324 HERA398332 HERA398330 HERA398329 HERA398328 HERA398327 HERA398325 HERA398326
Rekken voor diepvrieskasten met ultralage temperatuur van Revco
Rekken Rekken voor cryogene dozen
Maximise the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for Revco™ UxF and HERAfreeze™ HFU T freezers.
Bestelinformatie: Warranty varies by country, please contact us for details.
Leveringsinformatie: Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
Order Now
Specificaties
Environmentally Preferable