GC columns, J&W DB-5ms
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
121-5522 121-5542 122-5511 122-5512 122-5513 122-5516 122-5522 122-552A 122-5531 122-5532 122-5533 122-5536 122-5552 122-5561 122-5562 122-5563 123-5511 123-5512 123-5513 123-5526 123-5531 123-5532 123-5532E 123-5533 123-5536 123-5561 123-5562 123-5563 125-5512 125-5532 125-5537 125-553J 128-5512 128-5522 128-5552 122-5532G 122-5533E 122-5533G 122-5536G 122-5562G 123-5566 125-5537G 121-5522E 122-5532E 122-5536E 122-5562E 121-5522LTM 121-5523 121-5523LTM 122-5511LTM 122-5512E 122-5512LTM 122-5532LTM 122-5533LTM G3900-63004 123-5512LTM 123-5536LTM 125-553JLTM
HEWL121-5522EA 715 EUR
HEWL121-5522 HEWL121-5542 HEWL122-5511 HEWL122-5512 HEWL122-5513 HEWL122-5516 HEWL122-5522 HEWL122-552A 558-0365 558-0366 HEWL122-5533 HEWL122-5536 HEWL122-5552 HEWL122-5561 HEWL122-5562 HEWL122-5563 HEWL123-5511 HEWL123-5512 HEWL123-5513 HEWL123-5526 HEWL123-5531 HEWL123-5532 HEWL123-5532E HEWL123-5533 HEWL123-5536 HEWL123-5561 HEWL123-5562 HEWL123-5563 HEWL125-5512 HEWL125-5532 HEWL125-5537 HEWL125-553J HEWL128-5512 HEWL128-5522 HEWL128-5552 HEWL122-5532G HEWL122-5533E HEWL122-5533G HEWL122-5536G HEWL122-5562G HEWL123-5566 HEWL125-5537G HEWL121-5522E HEWL122-5532E HEWL122-5536E HEWL122-5562E HEWL121-5522LTM HEWL121-5523 HEWL121-5523LTM HEWL122-5511LTM HEWL122-5512E HEWL122-5512LTM HEWL122-5532LTM HEWL122-5533LTM HEWLG3900-63004 HEWL123-5512LTM HEWL123-5536LTM HEWL125-553JLTM
GC columns, J&W DB-5ms
Kolommen voor chromatografie
DB-5ms non-polar, low-bleed columns feature an improved signal to noise ratio for excellent sensitivity and mass spectral integrity of aromatic compounds.
  • Phenyl arylene polymer virtually equivalent to a (5% phenyl) methylpolysiloxane
  • Non-polar with very low bleed characteristics, ideal for GC/MS
  • Excellent inertness for active compounds
  • Improved signal to noise ratio for better sensitivity and mass spectral integrity
  • Bonded and cross-linked
J&W DB-5ms features an excellent signal to noise ratio for analytical applications. This improvedMore Product Information