Capillary columns, HP-PLOT MoleSieve
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
19091P-MS4 19091P-MS4E 19091P-MS7 19091P-MS8 19095P-MS0 19095P-MS0E 19095P-MS5 19095P-MS6 19095P-MS9 19095P-MS6E 19091P-MS8LTM 19095P-MS0LTM
HEWL19091P-MS4EA 870 EUR
HEWL19091P-MS4 HEWL19091P-MS4E HEWL19091P-MS7 HEWL19091P-MS8 HEWL19095P-MS0 HEWL19095P-MS0E HEWL19095P-MS5 HEWL19095P-MS6 HEWL19095P-MS9 HEWL19095P-MS6E HEWL19091P-MS8LTM HEWL19095P-MS0LTM
Capillary columns, HP-PLOT MoleSieve
Kolommen voor chromatografie
For applications requiring resolution of argon and oxygen without cryogenic cooling, select columns with thick film. The thin film configurations are ideal for many applications, including routine air monitoring and analysis. The thin film column resolves argon and oxygen at subambient temperatures.