Test standards for agilent J&W GC column
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
200-0010 200-0032 200-0070 200-0110 200-0113 200-0185 200-0310 200-0370
HEWL200-0010EA 69 EUR
HEWL200-0010 HEWL200-0032 HEWL200-0070 HEWL200-0110 HEWL200-0113 HEWL200-0185 HEWL200-0310 HEWL200-0370
Test standards for agilent J&W GC column
Standaarden Standaarden voor chromatografie
GC column test standards contains components that test the column for resolution characteristics, efficiency and inertness.
Leveringsinformatie: Test mixes are supplied at a concentration of 250 ng/µl in 2 ml vials.