Quartz cuvettes
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0100-1224 0100-1225 5061-3396 5061-3397 5061-3398 5062-2476 5063-6570 5063-6571 5063-6572 5063-6573 5065-9907 5065-9918 5188-8003 5188-8004 5188-8005
HEWL0100-1224EA 0 EUR
HEWL0100-1224 HEWL0100-1225 HEWL5061-3396 HEWL5061-3397 HEWL5061-3398 HEWL5062-2476 HEWL5063-6570 HEWL5063-6571 HEWL5063-6572 HEWL5063-6573 HEWL5065-9907 HEWL5065-9918 HEWL5188-8003 HEWL5188-8004 HEWL5188-8005
Quartz cuvettes
Cuvetten Cuvetten voor spectrofotometers
Flow cuvettes with center height of 15 mm and screw fitting connection.
For 8453 UV/Visible spectrophotometers.