Inlet septa
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5181-1260 5181-1261 5181-1283-100 5181-8815 5181-8816 5181-8833 5181-8839 5182-0853 5182-3442 5182-3444 5182-3445 5183-4757 5183-4758 5183-4759 5183-4760 5183-4761 5183-4762 5181-1283-50 5080-8894-100 5080-8728-100 5181-3383-50 5080-8896-50 5181-3383-100 5183-4759-100 5183-4761-100 5080-8728-50 5183-4757-100
HEWL5181-1260EA 46 EUR
HEWL5181-1260 HEWL5181-1261 HEWL5181-1283-100 HEWL5181-8815 HEWL5181-8816 HEWL5181-8833 HEWL5181-8839 HEWL5182-0853 HEWL5182-3442 HEWL5182-3444 HEWL5182-3445 HEWL5183-4757 HEWL5183-4758 HEWL5183-4759 HEWL5183-4760 HEWL5183-4761 HEWL5183-4762 HEWL5181-1283-50 HEWL5080-8894-100 HEWL5080-8728-100 HEWL5181-3383-50 HEWL5080-8896-50 HEWL5181-3383-100 HEWL5183-4759-100 HEWL5183-4761-100 HEWL5080-8728-50 HEWL5183-4757-100
Inlet septa
Tussenschoten
These septa are available for a variety of applications and have different upper temperature limits. The lower temperature septa are usually softer, seal better and can withstand more punctures (injections) than their high temperature counterparts.
  • Made up low bleed silicone rubber formulations
  • Efficient by using sharp syringe needles
  • Septa should be replaced to avoid leaks and sample loss
  • To be used with recommended temperature range